missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Equipment and Options for Erlab Enclosures and Safety Cabinets: This equipment completes and adapts Erlab ductless enclosures and microbiological safety cabinets to the specific requirements of each laboratory. Every option is designed and validated by Erlab to be integrated without compromising the containment and protection performance of the equipment.
- Designed and Validated by Erlab:
- Components engineered for a specific enclosure or cabinet reference
- Integration that preserves airflow, containment and operator protection
- Materials selected for chemical and biological resistance and easy decontamination
- Compatible with Erlab connected monitoring and safety programmes
- Adapt Your Workstation to Your Protocols: Extraction options, front window and access solutions, decontamination systems, sealing accessories and internal fittings allow the workstation to be configured for the products handled, the equipment installed inside the enclosure and the ergonomics required by the operator.
- Installation is designed to be carried out quickly, with no impact on the certification of the equipment when performed according to Erlab instructions.
- Key Benefits:
- Factory-validated option, fully compatible with the target equipment.
- Maintains containment and operator protection performance.
- Materials suitable for laboratory chemical and biological environments.
- Improves ergonomics, versatility and ease of use.
- Fast installation with no impact on equipment certification.
Specifications
Specifications
| Product Type | Enclosure |
| For Use With (Equipment) | Lois 900 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?