missing translation for 'onlineSavingsMsg'
Learn More

Clothiapine, TRC

Product Code. 30008129
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30008129 5 mg 5mg
30006405 10 mg 10mg
30007922 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30008129 Supplier Toronto Research Chemicals Supplier No. TRCC5885505MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2058-52-8
Molecular Formula C18 H18 Cl N3 S
Molecular Weight (g/mol) 343.87
Synonym 2-Chloro-11-(4-methyl-1-piperazinyl)dibenzo[b,f][1,4]thiazepine, 2-Chloro-11-(4-methylpiperazino)dibenzo[b,f][1,4]thiazepine, Clothiapin, Clothiapine, Clotiapine, Deliton, Entumine, Etumine, HF 2159, LW 2159, Dibenzo[b,f][1,4]thiazepine, 2-chloro-11-(4-methyl-1-piperazinyl)-
IUPAC Name 8-chloro-6-(4-methylpiperazin-1-yl)benzo[b][1,4]benzothiazepine
SMILES CN1CCN(CC1)C2=Nc3ccccc3Sc4ccc(Cl)cc24

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C18H18ClN3S/c1-21-8-10-22(11-9-21)18-14-12-13(19)6-7-16(14)23-17-5-3-2-4-15(17)20-18/h2-7,12H,8-11H2,1H3
Formula Weight 343.091
Percent Purity >95%
Chemical Name or Material Clothiapine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.