missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Clozapine N-oxide dihydrochloride

Product Code. 18776336
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18776336 10 mg 10mg
18796806 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18776336 Supplier R&D Systems™ Supplier No. 6329/10
  • ONLINE EXCLUSIVE PRICE
    224.06 €
    198.45 € / 10mg
    Save 25.61 €
    11% Off
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Activator of muscarinic DREADDs; water soluble version of Clozapine N-oxide (Cat. No. 4936)

Clozapine N-oxide dihydrochloride is a water-soluble salt of Clozapine N-oxide (Cat. No.4936). Activator of muscarinic DREADDs. Clozapine N-oxide dihydrochloride exhibits improved bioavailability (6 - 7-fold higher plasma concentration) compared with CNO, with less conversion to clozapine in animal studies.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 8-Chloro-11-(4-methyl-4-oxido-1-piperazinyl)-5 H-dibenzo[b,e][1,4 ]diazepine dihydrochloride
CAS 2250025-93-3
Assay Percent Range >99%
Quantity 10 mg
Product Line Tocris
Target DREADD Activators
Molecular Formula C18H19ClN4O0.2 HCl
Formula Weight Observed MW: 415.74
Purity 99%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.