missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ Clozapine
GREENER_CHOICE

Product Code. 18722784
Change view
Click to view available options
Quantity:
50 mg
500 mg
Unit Size:
500mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18722784 50 mg 50mg
30002209 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18722784 Supplier Tocris Bioscience™ Supplier No. 0444/50
  • ONLINE EXCLUSIVE PRICE
    239.00 €
    225.75 € / 50mg
    Save 13.25 €
    5% Off
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Dopamine antagonist with some D4 selectivity; also 5-HT2A/2C antagonist

Clozapine is an atypical antipsychotic drug, with a much lower tendency to cause extrapyramidal side effects than conventional neuroleptics. Displays a broad range of pharmacological actions; the antipsychotic effects are thought to be mediated principally by 5-HT2A/2C and dopamine receptor blockade (Ki values are 21, 170, 170, 230 and 330 nM for D4, D3, D1, D2 and D5 receptors respectively).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5786-21-0
Molecular Formula C18H19ClN4

Specifications

Quantity 50 mg
Formula Weight 326.83
Purity >99%
Molecular Weight (g/mol) 326.828
Chemical Name or Material Clozapine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.