missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ CLP 290

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
CLP 290 is a K+/Cl- Cotransporter 2 (KCC2) activator. Activates spinal inhibitory interneurons, restores physiological neuronal activity in the spinal cord and induces functional recovery in a mouse model of spinal cord injury. Also decreases blood vasopressin and glucose levels in mice with Streptozotocin (Cat. No. 1621) induced diabetes mellitus. Orally bioavailable prodrug of CLP 257 (Cat. No.5242).
Specifications
Specifications
| Chemical Name or Material | (Z)-5-Fluoro-2-((4-oxo-2-(tetrahydropyridazin-1(2 H)-yl)thiazol-5(4 H)-ylidene)methyl)phenyl pyrrolidine-1-carboxylate |
| CAS | 1181083-81-7 |
| Quantity | 10 mg |
| Target | KCC2 Activators |
| Molecular Formula | C19H21FN4O3S |
| Formula Weight | 1990 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction