missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Clusterin-like 1/CLUL1 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3185-SP
This item is not returnable.
View return policy
Description
Clusterin-like 1/CLUL1 Polyclonal specifically detects Clusterin-like 1/CLUL1 in Human samples. It is validated for Western Blot.Specifications
| Clusterin-like 1/CLUL1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLUL1, Clusterin like 1, clusterin-like 1 (retinal), clusterin-like protein 1, RA337M, Retinal-specific clusterin-like protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 27098 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Q15846 | |
| CLUL1 | |
| Mouse myeloma cell line NS0-derived recombinant human Clusterin-like 1/CLUL1 Cys20-Ala442 Accession # Q15846 | |
| 25 μg | |
| Primary | |
| Detects human Clusterin-like 1/CLUL1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 5% cross-reactivity with recombinant human Clusterin is observed. | |
| Human | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?