missing translation for 'onlineSavingsMsg'
Learn More

Emma Safety Footwear Clyde S3 Shoe

Product Code. 17592666
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
17592666 45 1 pair
17532666 39 1 pair
17542666 40 1 pair
17552666 41 1 pair
17562666 42 1 pair
17572666 43 1 pair
17582666 44 1 pair
17502676 46 1 pair
17512676 47 1 pair
17522676 48 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
This item is not returnable. View return policy
Product Code. 17592666 Supplier Emma Safety Footwear Supplier No. MM99656945
Add to basket
Add to basket
This item is not returnable. View return policy

If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clyde.

  • S3-Safetyshoe with sole plate for wet work
  • Metal Free
  • The streetborn safety shoe. If trainers are what you prefer to wear but you need a safer shoe for work, look no further than the Clyde.
  • A no compromise metal free safety shoe that looks and feels like a classic sneaker.
  • It gets its name from the highly supportive reinforced heel. This heel together with the shoe's reinforced, shock absorbing sole keeps ankles, shins and joints free from stress and injury.
  • DenimX laces with a story. Made from recycled jeans for our circular safety shoes.
  • Industries:
    • Logistics & transport
    • Electronics
    • Construction & technology
    • Manufacturing
    • Nature & environment

Specifications

Certifications/Compliance ISO 20345:2011
Closure Type Laces
Color Black
For Use With (Application) Logistics and Transport, Electronics, Construction and Technology, Manufacturing, Nature and Environment
Liner Material Hydro-Tec Antibacterial
Material Nubuck, Oiled Nubuck
Size 45
Fit Extra-wide Width
Ankle Style High
Product Type Safety Shoe
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.