Learn More
CM 11, Tocris Bioscience™
Homo-PROTAC™ for self-degradation of pVHL30
Brand: Tocris Bioscience 6416/5
Description
CM 11 is a PROTAC® for self-degradation of the long form of VHL, pVHL30 (DC50 < 100 nM). Composed of two VHL ligands joined by a linker. Exhibits > 1000-fold increased cellular activity compared to the parent VHL ligand, VH 032. Dimerizes pVHL30 with high avidity in vitro. Induced potent and rapid self-degradation of VHL in cell lines in a highly isoform dependent fashion. Negative control CMP 98 (Cat. No. 6417) also available.PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
| 2244684-49-7 | |
| N1,N20-bis((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)-3,6,9,12,15,18-hexaoxaicosanediamide | |
| 5 mg |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.