missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CM 11
GREENER_CHOICE

Product Code. 18781839
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
This item is not returnable. View return policy

Product Code. 18781839

Brand: Tocris Bioscience™ 6416/5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Homo-PROTAC™ for self-degradation of pVHL30

CM 11 is a PROTAC® for self-degradation of the long form of VHL, pVHL30 (DC50 < 100 nM). Composed of two VHL ligands joined by a linker. Exhibits > 1000-fold increased cellular activity compared to the parent VHL ligand, VH 032. Dimerizes pVHL30 with high avidity in vitro. Induced potent and rapid self-degradation of VHL in cell lines in a highly isoform dependent fashion. Negative control CMP 98 (Cat. No. 6417) also available.

PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.

TRUSTED_SUSTAINABILITY

Specifications

CAS 2244684-49-7
Synonym N1,N20-bis((S)-1-((2S,4R)-4-Hydroxy-2-((4-(4-methylthiazol-5-yl)benzyl)carbamoyl)pyrrolidin-1-yl)-3,3-dimethyl-1-oxobutan-2-yl)-3,6,9,12,15,18-hexaoxaicosanediamide
Quantity 5 mg
Molecular Formula C58H82N8O14S2
Formula Weight 1179.46
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.