Learn More
Skytec™ CMBH Boilersuit
Product Code. 31702836 Shop All Skytec Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
Unit Size:
Pack of 10
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size Unit Size
31702836 Medium Pack of 10
31702839 X-Small Pack of 10
31702835 Small Pack of 10
31702823 Large Pack of 10
31702827 X-Large Pack of 10
31702832 2X-Large Pack of 10
31702821 3X-Large Pack of 10
31702825 4X-Large Pack of 10
31702822 5X-Large Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 31702836 Supplier Skytec™ Supplier No. CCCMB/001300/GN2
  • ONLINE EXCLUSIVE PRICE
    1912.10 €
    1225.07 € / Pack of 10
    Save 687.03 €
    35% Off
Estimated Shipment: 07-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Chemmaster garments offer strong resistance suited for a wide range of chemical handling tasks.
  • Ring-pull zip for easy donning and doffing when wearing gloves
  • Double storm-facing stud closure for increased protection
  • Helmet-compatible hood
  • Raglan sleeve for greater freedom of movement
  • Garment can be reused if not contaminated or damaged
  • Stitched and high-frequency welded seams
  • Tested to ASTM F903
Specifications
Color Green
Hood Type Attached
Material PVC
Size Medium
Ankle Style Loose
Closure Type Zipper
Cuff Style Standard
Product Type Chemical Protective Clothing
Seam Type Welded