missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CMNB-Caged Carboxyfluorescein, SE (5-Carboxyfluorescein-Bis-(5-Carboxymethoxy-2-Nitrobenzyl) Ether, β-Alanine-Carboxamide, Succinimidyl Ester)
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Description
Conjugation of the succinimidyl ester of the water-soluble, CMNB-caged carboxyfluorescein, SE to a primary amine on a biomolecule of interest produces an essentially nonfluorescent probe that yields a green-fluorescent fluorescein-labeled product only after ultraviolet illumination and removal of the caging group.
Specifications
Specifications
| Quantity | 1 mg |
| Product Type | CMNB-Caged Carboxyfluorescein |
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Chemical Reactivity | Amine |
| Shipping Condition | Room Temperature |
| Reactive Moiety | Active Ester, Succinimidyl Ester |
| Label or Dye | FITC (Fluorescein) |
| Label Type | Classic Dyes |
For Research Use Only. Not for use in diagnostic procedures.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction