Learn More
Tocris Bioscience™ CMPD101
Potent and selective GRK2/3 inhibitor
Brand: Tocris Bioscience™ 5642/10
This item is not returnable.
View return policy
Description
CMPD101 is a potent and selective GRK2/3 inhibitor (IC50 values are 32 and 54 nM for GRK3 and GRK2, respectively). Exhibits selectivity for GRK2/3 over GRK1/5. Reduces DAMGO -induced desensitization and internalization of μ-opioid receptors.Specifications
| 3-[[[4-Methyl-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]methyl]amino]-N-[[2-(trifluoromethyl)phenyl]methyl]benzamide | |
| 865608-11-3 | |
| 10 mg |