missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CMPD101
GREENER_CHOICE

Product Code. 18744519
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18744519 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18744519 Supplier Tocris Bioscience™ Supplier No. 5642/10
  • ONLINE EXCLUSIVE PRICE
    249.00 €
    235.20 € / 10mg
    Save 13.80 €
    5% Off
 Restricted Item
Estimated Shipment: 31-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Potent and selective GRK2/3 inhibitor

CMPD101 is a potent and selective GRK2/3 inhibitor (IC50 values are 32 and 54 nM for GRK3 and GRK2, respectively). Exhibits selectivity for GRK2/3 over GRK1/5. Reduces DAMGO -induced desensitization and internalization of μ-opioid receptors.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-[[[4-Methyl-5-(4-pyridinyl)-4H-1,2,4-triazol-3-yl]methyl]amino]-N-[[2-(trifluoromethyl)phenyl]methyl]benzamide
CAS 865608-11-3
Quantity 10 mg
Target G Protein (Heterotrimeric) Inhibitors
Molecular Formula C24H21F3N6O
Formula Weight 466.46
Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.