Learn More
VELP Scientifica CN 802 Carbon Nitrogen Elemental Analyzer
Product Code. 30213536 Shop All Velp Products
Change view
Click to view available options
Type:
Elemental Analyzer
Unit Size:
Each
Product Code. Type Unit Size
30213536 Elemental Analyzer Each
1 options
Product Code. 30213536 Supplier VELP Scientifica Supplier No. F30800090
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
A fast, versatile, and highly sensitive elemental analyzer for simultaneous Carbon (Total and TOC) and Nitrogen determination ranging from micro to macro sample weights.
Fast, versatile, and highly sensitive elemental analyzer for simultaneous Carbon (Total and TOC) and Nitrogen determination ranging from micro to macro sample weights. The CN 802 offers high performance on both solid and liquid samples, working following international standards such as AOAC, AACC, ASBC, ISO, DIN, IFFO, OIV, ASTM, and EPA.
RAPID C/N DETERMINATION
  • The CN 802 performs analysis of both carbon and nitrogen in just 3-5 minutes.
  • The quick's start and sleeping mode ensures fast operation set-up with the possibility of leak test per zone
FLEXIBLE
  • Fully automatic determination of TC, TOC, TIC, TN and Carbon/Nitrogen Ratio on both solid and liquid samples.
  • Using dedicated accessories, it is possible to analyze liquid samples with high sensitivity and accuracy.
PRECISE
  • The HighSensIR Detector (Non-dispersive Infrared detector) and the LoGas™ TCD (Thermal Conductivity Detector) designed by VELP guarantee unmatched precision and unrivalled LOD.
EFFICIENT
  • The fully automatic and reliable combustion method of CN 802 together with the DriStep™ water trap provides reduced cost-per-analysis.
  • The 30-position disc autosampler expandable with an extra 3 discs ensures maximum productivity and uptime.
VERSATILE
  • The CN 802 works with solid, liquid and doughy samples of up to 1 gram.
  • Seamless carrier gas selection between Helium and Argon without hardware changes and with no memory effect.
SMART and SAFE
  • The CN 802 is easy to use thanks to the user-friendly CNSoft™ software which is equipped with maximum safety control of the instrument.
  • VELP Ermes cloud connection will deliver a new user experience and smart workflows
Specifications
Type Elemental Analyzer
Detector Type Thermal Conductivity Detector, HighSensIR
For Use With (Application) Environmental Science, Agronomy, Food Safety, Organic Chemistry