missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ CNQX

Product Code. 30001060
Change view
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30001060 50 mg 50mg
18014032 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30001060 Supplier R&D Systems™ Supplier No. 0190/50
  • ONLINE EXCLUSIVE PRICE
    914.66 €
    816.90 € / 50mg
    Save 97.76 €
    10% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Potent and selective non-NMDA iGluR antagonist

CNQX is an AMPA and kainate receptor antagonist (IC50 values are 0.3 μM and 1.5 μM for AMPA and kainate receptors, respectively). CNQX is also an antagonist at the glycine modulatory site on the NMDA receptor complex (IC50 = 25 μM). CNQX can be used to isolate GABAA receptor-mediated spontaneous inhibitory postsynaptic currents and antagonizes non-NMDA receptor-mediated responses in cultured cerebellar granule cells. CNQX shows neuroprotective effects in models of ischemia and inhibits seizure-like activity in hippocampal neurons.

CNQX Disodium Salt (Cat. No. 1045) also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115066-14-3
Molecular Formula C9H4N4O4

Specifications

Quantity 50 mg
Product Line Tocris
Formula Weight 232.16
Purity >99%
Molecular Weight (g/mol) 232.155
Chemical Name or Material CNQX
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.