missing translation for 'onlineSavingsMsg'
Learn More

BINDER™ CO2 Incubator Series CB

Product Code. 31741147 Shop All BINDER Products
Change view
Click to view available options
Dimensions (D x W x H) Interior:
330 x 400 x 400 mm
500 x 560 x 600 mm
575 x 620 x 750 mm
No. of Shelves:
2 std./3 max.
3 std./6 max.
3 std./8 max.
Model:
CB0170
CB056
CB170-L
CB170-O
CB170-OL
CB260
CB260-O
Unit Size:
Each
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (D x W x H) Interior No. of Shelves Model Unit Size
31741147 500 x 560 x 600 mm 3 std./6 max. CB170-L Each
31741123 330 x 400 x 400 mm 2 std./3 max. CB056 Each
31741129 330 x 400 x 400 mm 3 std./6 max. CB0170 Each
31741144 500 x 560 x 600 mm 3 std./6 max. CB170-O Each
31741117 500 x 560 x 600 mm 3 std./6 max. CB170-OL Each
31741114 575 x 620 x 750 mm 3 std./8 max. CB260 Each
31741112 575 x 620 x 750 mm 3 std./8 max. CB260-O Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
This item is not returnable. View return policy
Product Code. 31741147 Supplier BINDER™ Supplier No. 90400200
Powering Proteomics
Use promo code 28003
View all promotions
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Series CB - CO2 incubators with hot air sterilization and heat sterilizable CO2 sensor.

The powerful and versatile CB CO2 incubator is suitable for cell-based assays in drug and medical research. This BINDER incubator is even suitable for complex cultivation experiments or individual environments with hypoxic conditions thanks to its comprehensive assortment of options and accessories.

  • Safe - cell cultures receive maximum protection against contamination
  • Reliable - constant well-being conditions for the cells
  • Smart - simple routine cleaning and convenient operation
  • Economical - efficient operation without consumables

IMPORTANT FEATURES

  • Optimized humidification system with condensation protection
  • Tried-and-tested anti-contamination concept with 180°C hot air sterilization
  • Hot-air sterilizable CO2 sensor with single-beam infrared technology
  • Seamless inner chamber made of stainless steel with flanges as shelf support system
  • Fanless interior with Venturi CO2 gas mixing nozzle
  • Intuitive touchscreen controller
  • Internal data recording, can be read out in open format via USB interface
  • Fault diagnosis system with visual and acoustic alarm
  • Interfaces: Ethernet, USB, zero-voltage alarm contact
  • Fail-safe CO2 system for protecting the pH of cell cultures

OPTIONAL EQUIPMENT

  • Multi-sectional glass doors with special shelves for dividing up the space to accommodate different experiments
  • Stacking adapter for secure, space-saving installation
  • Gas tank changer for connecting two gas tanks to one or two incubators

Specifications

Controller Type MB 2
Capacity (Metric) 170 L
Chamber Material Stainless Steel (304/1.4301)
CO2 Concentration Range 0 to 20%
CO2 Sensor Technology IR Sensor
Convection Type Natural
Material Stainless Steel
No. of Shelves 3 std./6 max.
Plug Type EU
Relative Humidity 90 to 95%
Temperature Range (Metric) +4 above ambient to 50°C
Type CO2 Incubator
Display Yes
Model CB170-L
Temperature Uniformity 0.3 ± K
Dimensions (D x W x H) Interior 500 x 560 x 600 mm
Height (Metric) 870 mm
Depth (Metric) Interior 500 mm
Weight (Metric) 95 kg
Voltage 200/230 V
Frequency 50/60 Hz
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.