missing translation for 'onlineSavingsMsg'
Learn More

Coal tar, Thermo Scientific Chemicals

Product Code. 11399008
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11399008

Brand: Thermo Scientific Alfa Aesar 042488.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Coal tar is used for constructing roads, electricity generation and coking. It has been used as topical medical treatment for psoriasis and dandruff and is formulated as a soap or ointment. It is also used for other types of rashes (such as eczema, atopic dermatitis, chronic exudative dermatitis).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 8007-45-2
MDL Number MFCD00146482
Synonym Tar

Specifications

Odor Petroleum-like
Quantity 100 mL
Merck Index 14,2426
Solubility Information Not miscible or difficult to mix with water.
Physical Form Viscous Liquid
Chemical Name or Material Coal tar

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.