Learn More
BINDER™ Coated Heat Exchanger
Product Code. 30238286 Shop All BINDER Products
Change view
Click to view available options
For Use With (Equipment):
KB PRO 1600
KB PRO 260
KB PRO 470
KB PRO 720, KB PRO 1060
KBF PRO 130
KBF PRO 1600
KBF PRO 260
KBF PRO 470
KBF PRO 720, KBF PRO 1060
Unit Size:
Each
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
30238286 KB PRO 260 Each
30238248 KB PRO 470 Each
30238208 KB PRO 720, KB PRO 1060 Each
30238303 KBF PRO 130 Each
30238380 KB PRO 1600 Each
30238376 KBF PRO 260 Each
30238318 KBF PRO 470 Each
30238329 KBF PRO 720, KBF PRO 1060 Each
30238287 KBF PRO 1600 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 30238286 Supplier BINDER™ Supplier No. 80122462
Heat exchanger that is coated in plastic to prevent damage.
Heat exchanger that is coated in plastic to prevent damage to the heat exchanger caused by corrosive sample material.
Specifications
For Use With (Equipment) KB PRO 260
Material Aluminum
Type Heat Exchanger