missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ ELISA Phosphate Coating Buffer

Product Code. 10592615
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10592615 100 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10592615 Supplier Thermo Scientific™ Supplier No. CB07100
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA.

  • ELISA Phosphate Coating Buffer is intended for use as a plate coating buffer for Invitrogen Antibody Pairs or when developing a sandwich ELISA.
  • The buffer is a 10 mM phosphate buffer, pH 7.4.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Coating Buffer
Quantity 100 mL
Conjugate Enzyme Labeled
Form Concentrate
For Use With (Application) ELISA
Content And Storage Buffer. Store at RT.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.