missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Carbonyl (Stabilized with 1-5% Hexane), TRC

Product Code. 30352361
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352361 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352361 Supplier Toronto Research Chemicals Supplier No. TRCC633100250MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cobalt Carbonyl
CAS 10210-68-1
Molecular Formula C8Co2O8
Quantity 250 mg
Synonym Cobalt octacarbonyl (Co2(CO)8), Cobalt Tetracarbonyl Dimer, Di-μ-carbonylhexacarbonyldicobalt, Dicobalt Carbonyl, Dicobalt Octacarbonyl, Octacarbonyldicobalt
InChI Formula InChI=1S/8CO.2Co/c8*1-2;;/q;;;;;;;;2*+2
SMILES [Co+2].[Co+2].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+]
IUPAC Name carbon monoxide;cobalt(2+)
Molecular Weight (g/mol) 341.95
Formula Weight 341.8257
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.