missing translation for 'onlineSavingsMsg'
Learn More

Cobalt foil, 0.1mm (0.004in) thick, 99.95% (metals basis)

Product Code. 11375377
Change view
Click to view available options
Quantity:
25 x 25 mm
50 x 50 mm
100 x 100 mm
Unit Size:
25mm
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11375377 100 x 100 mm Each
11385377 25 x 25 mm 25mm
11356129 50 x 50 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11375377 Supplier Thermo Scientific Alfa Aesar Supplier No. 042658.GH
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Form: Approx.0.56g/25 x 25mm | CAS: 7440-48-4 | Co | 58.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-48-4
Molecular Formula Co
Molecular Weight (g/mol) 58.93
MDL Number MFCD00010935
InChI Key GUTLYIVDDKVIGB-UHFFFAOYSA-N
Synonym kobalt, cobalto, moncation, co1+, cobaltum, powder, cobalt, ion co1+, hydrido, atom, hydride
PubChem CID 104730
ChEBI CHEBI:27638
IUPAC Name cobalt
SMILES [Co]

Specifications

Color Gray
Appearance Gray foil
Density 8.92g/cm3
Quantity 100 x 100 mm
Odor Odorless
Assay Percent Range 99.95% (Metals basis)
Form Foil
Chemical Name or Material Cobalt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.