missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) 2-methoxyethoxide, 99% (metals basis), 5% w/v in 2-methoxyethanol

Product Code. 11379597
Change view
Click to view available options
Quantity:
25 mL
100 mL
Unit Size:
100mL
25mL
Product Code. Quantity unitSize
11379597 25 mL 25mL
11369597 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 11379597

Brand: Thermo Scientific Alfa Aesar 041237.AC

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 142600-61-1 | C6H14CoO4 | 209.107 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 142600-61-1
Molecular Formula C6H14CoO4
Molecular Weight (g/mol) 209.107
MDL Number MFCD00798554
InChI Key BFBNUELICQBJBC-UHFFFAOYSA-N
Synonym cobalt ii 2-methoxyethoxide
PubChem CID 131873636
IUPAC Name cobalt(2+);2-methoxyethanolate
SMILES COCC[O-].COCC[O-].[Co+2]

Specifications

Color Brown
Density 0.96 g/mL
Boiling Point 124°C
Flash Point 40°C (104°F)
Linear Formula Co(OCH2CH2OCH3)2
Quantity 25 mL
UN Number UN1188
Sensitivity Moisture sensitive
Formula Weight 209.11
Percent Purity 99%
Concentration or Composition (by Analyte or Components) 5% w/v in 2-methoxyethanol
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Cobalt(II) 2-methoxyethoxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.