missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Acetate Tetrahydrate, 98.0 to 102.0%, ACS Reagent, Honeywell™ Fluka™

Product Code. 15615430 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15615430 100 g Poly bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15615430 Supplier Honeywell-Fluka Supplier No. 403024100G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥98.0%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

Melting Point 298°C
CAS 6147-53-1
Physical Form Crystalline
Quantity 100 g
Molecular Formula C4H14CoO8
Linear Formula (CH3COO)2Co · 4H2O
MDL Number MFCD00149569
UN Number UN3077
Beilstein 3731397
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
IUPAC Name cobalt(2+);diacetate;tetrahydrate
Molecular Weight (g/mol) 249.081
PubChem CID 61372
Formula Weight 249.08g/mol
Percent Purity 98.0 to 102.0%
Packaging Poly bottle
Chemical Name or Material Cobalt(II) acetate tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.