missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate tetrahydrate, 98%

Product Code. 11334549
Change view
Click to view available options
Quantity:
25 g
50 g
100 g
500 g
Unit Size:
100g
25g
500g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11334549 100 g 100g
11324549 25 g 25g
11468297 50 g 50g
11344549 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11334549 Supplier Thermo Scientific Alfa Aesar Supplier No. A18374.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol

Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

Solubility
Soluble in water, alcohol, dilute acids and phenyl acetate.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372

Specifications

Melting Point ∼140°C (in -4H2O)
Density 1.71
Odor Odorless
Linear Formula Co(OOCCH3)2·4H2O
Refractive Index 1.542
Quantity 100 g
UN Number UN3077
Beilstein 3731397
Sensitivity Hygroscopic
Merck Index 14,2433
Solubility Information Soluble in water,alcohol,dilute acids and phenyl acetate.
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]
IUPAC Name cobalt(2+);diacetate;tetrahydrate
Molecular Weight (g/mol) 249.081
Formula Weight 249.08 (177.02 Anhydrous)
Percent Purity 98%
Chemical Name or Material Cobalt(II) acetate tetrahydrate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.