missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate tetrahydrate, 98%

Product Code. 11478297
Change view
Click to view available options
Quantity:
250 g
Unit Size:
250g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11478297 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11478297 Supplier Thermo Scientific Alfa Aesar Supplier No. A18374.30
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6147-53-1 | C4H14CoO8 | 249.08 g/mol

Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

Solubility
Soluble in water, alcohol, dilute acids and phenyl acetate.

Notes
Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 6147-53-1
Melting Point ∼140°C (in -4 H2O)
Density 1.71
Odor Odorless
Molecular Formula C4H14CoO8
Linear Formula Co(OOCCH3)2·4 H2O
Refractive Index 1.542
MDL Number MFCD00149569
Quantity 250 g
UN Number UN3077
Beilstein 3731397
Sensitivity Hygroscopic
Merck Index 14,2433
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt
Solubility Information Soluble aq. soln.,alcohol,dilute acids and phenyl acetate.
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILES O.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
IUPAC Name cobalt(2+);diacetate;tetrahydrate
Molecular Weight (g/mol) 249.08
PubChem CID 61372
Formula Weight 249.08 (177.02 Anhydrous)
Percent Purity 98%
Chemical Name or Material Cobalt(II) acetate tetrahydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) acetate tetrahydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contain: Acetic acid, cobalt(2+) salt, tetrahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.