missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate tetrahydrate, ACS, 98.0-102.0%

Product Code. 11478470
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11478470 25 g 25g
11468470 100 g 100g
11488470 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11478470 Supplier Thermo Scientific Alfa Aesar Supplier No. 044344.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol

Cobalt(II) acetate tetrahydrate is used as an industrial catalyst to harden paints and varnishes, an active catalyst for oxidation and esterification reactions. It is also used to prepare metal complexes having unusual coordination geometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 6147-53-1
Molecular Formula C4H14CoO8
Molecular Weight (g/mol) 249.081
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
IUPAC Name cobalt(2+);diacetate;tetrahydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]

Especificaciones

Melting Point ∼140°C (in -4H2O)
Color Red
Physical Form Crystalline
Quantity 25 g
Linear Formula Co(OOCCH3)2·4H2O
UN Number UN3077
Beilstein 3731397
Merck Index 14,2433
Solubility Information Soluble in water,alcohol,dilute acids and phenyl acetate.
Formula Weight 249.08 (177.02 Anhydrous)
Percent Purity 98.0 to 102.0%
Grade ACS
Odor Odorless
Refractive Index 1.542
Sensitivity Hygroscopic
Density 1.71 g/mL
Chemical Name or Material Cobalt (II) acetate tetrahydrate
Mostrar más Mostrar menos

RUO – Research Use Only

Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.