missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) acetate tetrahydrate, Co 24%

Product Code. 11336648
Change view
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11336648 1 kg 1kg
11326648 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11336648 Supplier Thermo Scientific Alfa Aesar Supplier No. 012227.A1
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 6147-53-1 | C4H14CoO8 | 249.081 g/mol

It is used as an industrial catalyst that allow paints and varnishes to harden. Also as a precursor to various oil drying agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
Molecular Weight (g/mol) 249.081
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
IUPAC Name cobalt(2+);diacetate;tetrahydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]

Specifications

Melting Point ∽140°C-4H2O
Color Red
Physical Form Crystals
Quantity 1 kg
Assay Percent Range 24%
Beilstein 3731397
Solubility Information Soluble in water,alcohol,dilute acids
Formula Weight 249.08 (177.02 Anhydrous)
Odor Odorless
Refractive Index 1.542
Sensitivity Hygroscopic
Density 1.71
Chemical Name or Material Cobalt(II) acetate tetrahydrate, Co 24%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.