missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Acetate Tetrahydrate, Pure, Fisher Chemical

Product Code. 10315460 Shop All Fisher Chemical Products
Change view
Click to view available options
250g, Plastic powder jar
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10315460 250g, Plastic powder jar
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10315460 Supplier Fisher Chemical Supplier No. C/6400/50
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Melting Point 140°C
CAS 6147-53-1
Color Red
pH 6.8
Physical Form Solid
Quantity 250 g
Molecular Formula C4H14CoO8
MDL Number 149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
SMILES O.O.O.O.[Co++].CC([O-])=O.CC([O-])=O
IUPAC Name λ²-cobalt(2+) diacetate tetrahydrate
Molecular Weight (g/mol) 249.08
Formula Weight 249.08g/mol
Packaging Plastic powder jar
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.