missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Acetate Tetrahydrate, Reagent grade, Honeywell™

Product Code. 15621840 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15621840 50 g Poly Bottle 50g
15611840 250 g Poly Bottle 250g
15601840 1 kg Poly Bottle 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15621840 Supplier Honeywell Research Chemicals Supplier No. 20839650G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6147-53-1
Molecular Formula C4H14CoO8
Molecular Weight (g/mol) 249.081
MDL Number MFCD00149569
InChI Key ZBYYWKJVSFHYJL-UHFFFAOYSA-L
Synonym cobalt ii acetate tetrahydrate, cobalt acetate tetrahydrate, cobaltous acetate tetrahydrate, cobalt diacetate tetrahydrate, bis acetato tetraquacobalt, octan kobaltnaty czech, acetic acid, cobalt 2+ salt, tetrahydrate, octan kobaltnaty, ccris 9441
PubChem CID 61372
IUPAC Name cobalt(2+);diacetate;tetrahydrate
SMILES CC(=O)[O-].CC(=O)[O-].O.O.O.O.[Co+2]

Specifications

Melting Point 298°C
Quantity 50 g
Linear Formula (CH3COO)2Co · 4H2O
UN Number UN3077
Beilstein 3731397
Formula Weight 249.08g/mol
Grade Reagent
Packaging Poly Bottle
Appearance Crystals
Density 1.705g/cu.cm
Chemical Name or Material Cobalt(Ii) Acetate Tetrahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.