missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Carbonate Hydrate, Honeywell™ Fluka™

Product Code. 15631740 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Poly Bottle
Unit Size:
1kg
250g
50g
Product Code. Quantity Packaging unitSize
15631740 50 g Poly Bottle 50g
15621740 250 g Poly Bottle 250g
15611740 1 kg Poly Bottle 1kg
3 options
This item is not returnable. View return policy

Product Code. 15631740

Brand: Honeywell-Fluka 20219350G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 57454-67-8
Molecular Formula CCoO3
Molecular Weight (g/mol) 118.94
MDL Number MFCD00149654
InChI Key ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
Synonym cobalt ii carbonate hydrate, cobalt carbonate hydrate, acmc-20aqp2, co3.co.h2o, ksc169q7l, cobalt ii carbonate hydrate-co co min
PubChem CID 16211458
IUPAC Name λ²-cobalt(2+) carbonate
SMILES [Co++].[O-]C([O-])=O

Specifications

Quantity 50 g
Linear Formula CoCO3·xH2O
UN Number NONH for all modes of transport
Formula Weight 118.94g/mol (anhydrous basis)
Packaging Poly Bottle
Density 4.13g/mL at 25°C
Chemical Name or Material Cobalt(Ii) Carbonate Hydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.