missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) carbonate hydrate

Product Code. p-46406 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
Packaging:
Plastic bottle
Unit Size:
1kg
250g
50g
This item is not returnable. View return policy

Product Code. 10022590

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 57454-67-8 | CCoO3 | 118.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57454-67-8
Molecular Formula CCoO3
Molecular Weight (g/mol) 118.94
MDL Number MFCD00149654
InChI Key ZOTKGJBKKKVBJZ-UHFFFAOYSA-L
Synonym cobalt ii carbonate hydrate, cobalt carbonate hydrate, acmc-20aqp2, co3.co.h2o, ksc169q7l, cobalt ii carbonate hydrate-co co min
PubChem CID 16211458
SMILES [Co++].[O-]C([O-])=O

Specifications

Color White to Yellow
Physical Form Mass, Crystals, Crystalline Powder and/or Chunks
Quantity 50 g
Linear Formula CoCO3·xH2O
Merck Index 14,2436
Solubility Information Solubility in water: insoluble
IUPAC Name λ²-cobalt(2+) carbonate
Formula Weight 118.94
Percent Purity ≥45.5% (Cobalt) (Complexometry)
Packaging Plastic bottle
Chemical Name or Material Cobalt(II) carbonate hydrate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) carbonate hydrate
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H341-Suspected of causing genetic defects.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.