missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride, 97%, anhydrous

Product Code. 10677162 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10677162 5 g Glass Bottle 5g
10593761 100 g Plastic bottle 100g
10656982 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10677162 Supplier Thermo Scientific Acros Supplier No. 214130050
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 7646-79-9 | Cl2Co | 129.83 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-79-9
Molecular Formula Cl2Co
Molecular Weight (g/mol) 129.83
MDL Number MFCD00010938
InChI Key GVPFVAHMJGGAJG-UHFFFAOYSA-L
Synonym cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride
IUPAC Name λ²-cobalt(2+) dichloride
SMILES [Cl-].[Cl-].[Co++]

Specifications

Boiling Point 1049.0°C
Melting Point 735.0°C
Color Blue
Physical Form Powder
Quantity 5 g
Assay Percent Range 97%
Linear Formula CoCl2
Fieser 01,155; 06,127; 08,291; 10,101; 13,249; 14,99; 15,97
Merck Index 15, 2421
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol and acetone
Formula Weight 129.84
Percent Purity 97%
Packaging Glass Bottle
Density 3.35
Chemical Name or Material Cobalt(II) chloride, 97%
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) chloride, anhydrous
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Serious eye damage/eye irritation Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H341-Suspected of causing genetic defects.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.