missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Chloride, 97%, Honeywell Fluka™

Product Code. 15682310 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
Packaging:
Glass Bottle
Unit Size:
100g
500g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15682310 5 g Glass Bottle 5g
15662310 100 g Glass Bottle 100g
15672310 500 g Glass Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15682310 Supplier Honeywell-Fluka Supplier No. 2326965G

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

0.97

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-79-9
Molecular Formula Cl2Co
Molecular Weight (g/mol) 129.83
MDL Number MFCD00010938
InChI Key GVPFVAHMJGGAJG-UHFFFAOYSA-L
Synonym cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride
IUPAC Name λ²-cobalt(2+) dichloride
SMILES [Cl-].[Cl-].[Co++]

Specifications

Boiling Point 1049°C
Melting Point 724°C
Color Blue
Quantity 5 g
Linear Formula CoCl2
UN Number UN3077
Formula Weight 129.84g/mol
Percent Purity 97%
Packaging Glass Bottle
Appearance Powder
Vapor Pressure 40mmHg (0°C)
Density 3.37g/cm3
Chemical Name or Material Cobalt(Ii) Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.