missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) chloride, anhydrous, 97%

Product Code. 11459517
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11459517 500 g 500g
11439517 25 g 25g
11449517 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11459517 Supplier Thermo Scientific Alfa Aesar Supplier No. B22031.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7646-79-9 | Cl2Co | 129.83 g/mol

Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cobalt(II) chloride is used in humidity indicator in weather instruments. In the anhydrous form, it finds use in electroplating of cobalt, in organic chemistry and is a precursor to cobaltocene, (bis(cyclopentadienyl)cobalt(II), which is a good reducing agent. It also serves as a Lewis acid. Cobalt chloride is an indicator for water in desiccants, owing to the reversible hydration/dehydration coupled with distinct color change. Cobalt chloride is useful for producing invisible ink as it turns blue when heated and becomes invisible once it gets cooled. Cobalt(II) chloride catalyzes cross coupling of aryl halides or vinyl halides with aryl grignard reagents in excellent yields.

Solubility
Freely soluble in water and methanol. Soluble in alcohol, acetone, ether, glycerol, and pyridine.

Notes
Hygroscopic. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7646-79-9
Molecular Formula Cl2Co
Molecular Weight (g/mol) 129.83
MDL Number MFCD00010938
InChI Key GVPFVAHMJGGAJG-UHFFFAOYSA-L
Synonym cobalt ii chloride, cobaltchloride, cobalt chloride 0.1 m solution, cobalt ii chloride, anhydrous, cobalt ii-chloride, curator_000013, cobalt chloride anhydrous, wln: co g2, cobalt-ii-chloride
IUPAC Name λ²-cobalt(2+) dichloride
SMILES [Cl-].[Cl-].[Co++]

Specifications

Boiling Point 1049°C
Melting Point 735°C
Quantity 500 g
Linear Formula CoCl2
UN Number UN3077
Merck Index 14,2437
Solubility Information Freely soluble in water and methanol. Soluble in alcohol,acetone,ether,glycerol,and pyridine.
Formula Weight 129.84
Percent Purity 97%
Odor Odorless
Sensitivity Hygroscopic
Density 3.367
Chemical Name or Material Cobalt(II) chloride, anhydrous
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.