missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Chloride Hexahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10716671 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
Product Code. Quantity unitSize
10716671 100 g 100g
10273610 500 g 500g
10375550 1 kg 1kg
12668625 5 kg 5kg
4 options
This item is not returnable. View return policy

Product Code. 10716671

Brand: Fisher Chemical C/6560/48

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 7791-13-1
Molecular Formula Cl2CoH12O6
Molecular Weight (g/mol) 237.92
MDL Number MFCD00149652
InChI Key GFHNAMRJFCEERV-UHFFFAOYSA-L
IUPAC Name λ²-cobalt(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Cl-].[Cl-].[Co++]

Specifications

Melting Point 86°C
Color Red to Violet
pH 4.6
Physical Form Crystalline Solid
Quantity 100 g
Formula Weight 237.93g/mol
Packaging Plastic powder jar
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.