missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) gluconate hydrate

Product Code. 11475707
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11475707 25 g 25g
11485707 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11475707 Supplier Thermo Scientific Alfa Aesar Supplier No. B24293.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1809162-99-9 | C12H26CoO15 | 469.258 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1809162-99-9
Molecular Formula C12H26CoO15
Molecular Weight (g/mol) 469.258
MDL Number MFCD02684006
InChI Key QBVYGKYWWCNZSO-XRDLMGPZSA-N
Synonym cobalt ii gluconate hydrate, 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoyl oxy cobaltio 2r,3s,4r,5r-2,3,4,5,6-pentahydroxyhexanoate hydrate
PubChem CID 117064710
IUPAC Name cobalt;(2R,3S,4R,5R)-2,3,4,5,6-pentahydroxyhexanoic acid;hydrate
SMILES C(C(C(C(C(C(=O)O)O)O)O)O)O.C(C(C(C(C(C(=O)O)O)O)O)O)O.O.[Co]

Specifications

Melting Point >300°C
Quantity 25 g
Formula Weight 449.23 (Anhydrous)
Chemical Name or Material Cobalt(II) gluconate hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.