missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Nitrate Hexahydrate, 98%, Reagent grade, Honeywell™

Product Code. 15622290 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
6 x 500 g
Packaging:
Poly Bottle
Unit Size:
100g
500g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15622290 6 x 500 g Poly Bottle Pack of 6
15602290 100 g Poly Bottle 100g
15612290 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15622290 Supplier Honeywell Research Chemicals Supplier No. 2303756X500G
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Reagent Grade, 98%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
Synonym cobalt nitrate hexahydrate, cobalt ii nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate
PubChem CID 24821
ChEBI CHEBI:86214
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C
Color Red
Quantity 6 x 500 g
Linear Formula Co(NO3)2 · 6H2O
UN Number UN1477
IUPAC Name cobalt(2+);dinitrate;hexahydrate
Formula Weight 291.03g/mol
Percent Purity 98%
Grade Reagent
Packaging Poly Bottle
Appearance Crystals or chips
Density 1.87g/cm3
Chemical Name or Material Cobalt(Ii) Nitrate Hexahydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.