missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For analysis
Specifications
Specifications
| Melting Point | 55.0°C |
| Color | Brown to Red |
| Physical Form | Crystalline Powder or Chips |
| Quantity | 500 g |
| Assay Percent Range | 99% min. (Complexometry) |
| Linear Formula | Co(NO3)2·6H2O |
| Merck Index | 15, 2428 |
| Solubility Information | Solubility in water: 1340g/L. Other solubilities: soluble in ethanol and acetone, soluble in most common organic solvents |
| Formula Weight | 291.02 |
| Percent Purity | ≥99% |
| Show More |
Safety and Handling
Product Identifier
- Cobalt(II) nitrate hexahydrate
- Danger
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Oxidising solid Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- H272-May intensify fire; oxidiser.
- H302-Harmful if swallowed.
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H332-Harmful if inhaled.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H360F-May damage fertility.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- MIXTURE LIST-Contains : Cobalt(II) nitrate,hexahydrate
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?