missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Nitrate Hexahydrate, Certified AR for Analysis, Fisher Chemical™

Product Code. 10529380 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10529380 100 g 100g
10000090 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10529380 Supplier Fisher Chemical Supplier No. C/6680/48
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Specifications

Melting Point 55°C
Color Brown-Red
Physical Form Solid
Quantity 100 g
Formula Weight 291.02g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) nitrate hexahydrate
Signal Word
  • Danger
Hazard Category
  • Oxidising solid Category 2
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Acute toxicity Category 4
  • Skin sensitiser Category 1
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 1B
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contain: Cobalt nitrate hexahydrate, Cobalt(II) nitrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.