missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical™

Product Code. 10634912 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10634912 500 g 500g
10080080 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10634912 Supplier Fisher Chemical Supplier No. C/6640/53
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

Este artículo no se puede devolver. Vea la política de devoluciones

Identificadores de productos químicos

CAS 10026-22-9
Molecular Formula CoH12N2O12
Molecular Weight (g/mol) 291.03
MDL Number MFCD00149647
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
IUPAC Name λ²-cobalt(2+) hexahydrate dinitrate
SMILES O.O.O.O.O.O.[Co++].[O-][N+]([O-])=O.[O-][N+]([O-])=O

Especificaciones

Melting Point 55°C
Color Brown-Red
Physical Form Solid
Quantity 500 g
Formula Weight 291.02g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons compliance-icons compliance-icons compliance-icons
Nombre del producto
  • Cobalt(II) nitrate hexahydrate
Palabra de advertencia
  • Danger
Clasificación
  • Oxidising solid Category 2
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Indicaciones de peligro
  • H272-May intensify fire; oxidiser.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
Consejos de prudencia
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.