missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) perchlorate hexahydrate, 99.999% (metals basis)

Product Code. 11448430
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11448430 50 g 50g
11438430 10 g 10g
11458430 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11448430

Brand: Thermo Scientific Alfa Aesar 044320.18

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13478-33-6 | Cl2CoO8 | 257.825 g/mol

It is used as a chemical and organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13478-33-6
Molecular Formula Cl2CoO8
Molecular Weight (g/mol) 257.825
MDL Number MFCD00150207
InChI Key BSUSEPIPTZNHMN-UHFFFAOYSA-L
Synonym cobaltous perchlorate, cobalt diperchlorate, cobalt ii perchlorate, cobalt diperchlorate hexahydrate, cobalt perchlorate hexahydrate, cobaltous perchlorate, hexahydrate, cobalt ii perchlorate, hexahydrate, perchloric acid, cobalt ii salt, hexahydrate, diperchloratocobalt
PubChem CID 26064
IUPAC Name cobalt(2+);diperchlorate
SMILES [O-]Cl(=O)(=O)=O.[O-]Cl(=O)(=O)=O.[Co+2]

Specifications

Physical Form Crystalline
Quantity 50 g
UN Number UN1481
Solubility Information Soluble in water.
Formula Weight 366.74 (257.83 Anhydrous)
Percent Purity 99.999%
Odor Odorless
Sensitivity Hygroscopic
Assay (metals basis)
Chemical Name or Material Cobalt (II) perchlorate hexahydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.