missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) phthalocyanine

Product Code. 11384238
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11384238 5 g 5g
11374238 1 g 1g
11364238 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11384238 Supplier Thermo Scientific Alfa Aesar Supplier No. 041496.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 3317-67-7 | C32H16CoN8 | 571.469 g/mol

Cobalt(II) phthalocyanine may be used to develop carbon black based electrocatalysts. CoPC may be used as an end capping agent of a hyperbranched poly(aryl ether ketone) to be used for oxidative decomposition of 2,4,6,-trichlorophenol. CoPC modified carbon paste may be used as indicators for electrocatalytic amperometric measurements. ZnO impregnated with CoPC may be used as a sensitizer to determine the photodegradation of cyanide in aqueous suspension. CoPC catalyzed aerobic regenerations of aldehydes and ketones have been investigated.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3317-67-7
Molecular Formula C32H16CoN8
Molecular Weight (g/mol) 571.469
MDL Number MFCD00010718
InChI Key MPMSMUBQXQALQI-UHFFFAOYSA-N
Synonym cobalt ii phthalocyanine, cobalt phthalocyanine, phthalocyanine cobalt ii, phthalocyanine, cobalt, cobalt ii phthalocyanine, beta-form, dye content 97 %
PubChem CID 2734991
SMILES C1=CC=C2C(=C1)C3=NC4=NC(=NC5=C6C=CC=CC6=C([N-]5)N=C7C8=CC=CC=C8C(=N7)N=C2[N-]3)C9=CC=CC=C94.[Co+2]

Specifications

Color Blue to Purple
Odor Odorless
Quantity 5 g
Solubility Information Insoluble in water.
Formula Weight 571.47
Physical Form Powder
Chemical Name or Material Cobalt(II) phthalocyanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.