missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate heptahydrate, for analysis

Product Code. 15234318 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 g
1 kg
Unit Size:
1kg
250g
This item is not returnable. View return policy

Product Code. 15234318

Brand: Thermo Scientific Acros 453902500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 10026-24-1 | CoH14O11S | 281.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
PubChem CID 61444
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

Melting Point 98.0°C
pH 4 to 6 (5% aq. soln.)
Quantity 250 g
Linear Formula CoSO4·7H2O
Merck Index 15,2432
Solubility Information Solubility in water: 362g/L (20°C). Other solubilities: slightly soluble in methanol and ethanol
IUPAC Name cobalt(2+);sulfate;heptahydrate
Formula Weight 281.09
Percent Purity 99.0 to 101.0% (Complexometry)
Grade Analytical
Chemical Name or Material Cobalt(II) sulfate heptahydrate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cobalt(II) sulfate heptahydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Carcinogenicity Category 1B
  • Germ cell mutagenicity Category 2
  • Reproductive toxicity Category 1B
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H302-Harmful if swallowed.
  • H317-May cause an allergic skin reaction.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H341-Suspected of causing genetic defects.
  • H350i-May cause cancer by inhalation.
  • H360F-May damage fertility.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
Supplemental information
  • MIXTURE LIST-Contains : Cobalt(II) sulfate heptahydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.