missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) sulfate heptahydrate, Honeywell™

Product Code. 15607770 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 g
250 g
1 kg
2.5 kg
Unit Size:
100g
1kg
2.5kg
250g
This item is not returnable. View return policy

Product Code. 15607770

Brand: Honeywell Research Chemicals C6768100G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-24-1
Molecular Formula CoH14O11S
Molecular Weight (g/mol) 281.09
MDL Number MFCD00149658
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
PubChem CID 61444
IUPAC Name cobalt(2+);sulfate;heptahydrate
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O

Specifications

Quantity 100 g
Linear Formula CoSO4 · 7H2O
UN Number UN3077
Formula Weight 281.1g/mol
Percent Purity ≥99%
Density 2.03g/mL at 25°C
Chemical Name or Material Cobalt(II) sulfate heptahydrate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.