missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(Ii) Sulfate Heptahydrate, Solstice

Product Code. 15641230 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15641230 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15641230 Supplier Honeywell-Fluka Supplier No. 12933500G
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

Este artículo no se puede devolver. Vea la política de devoluciones

Purum,≥95%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Especificaciones

CAS 10026-24-1
Quantity 500 g
Molecular Formula CoH14O11S
Linear Formula CoSO4 · 7H2O
MDL Number MFCD00149658
UN Number 3077
Synonym cobalt sulfate heptahydrate, cobalt 2+ sulfate heptahydrate, cobaltous sulfate heptahydrate, cobalt ii sulfate heptahydrate, cobalt monosulfate heptahydrate, dsstox_cid_340, dsstox_rid_75523, dsstox_gsid_20340, bieberite, coo4s.7h2o
InChI Key MEYVLGVRTYSQHI-UHFFFAOYSA-L
SMILES O.O.O.O.O.O.O.[Co++].[O-]S([O-])(=O)=O
IUPAC Name cobalt(2+);sulfate;heptahydrate
Molecular Weight (g/mol) 281.09
PubChem CID 61444
Formula Weight 281.1g/mol
Percent Purity ≥95%
Grade purum
Density 2.03g/mL at 25°C
Chemical Name or Material Cobalt(Ii) Sulfate Heptahydrate
Mostrar más Mostrar menos
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.