missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cobalt(II) Thiocyanate, TRC
Click to view available options
Quantity:
5 g
Unit Size:
5g
Specifications
Specifications
| Chemical Name or Material | Cobalt(II) Thiocyanate |
| CAS | 3017-60-5 |
| Molecular Formula | Co (S C N)2 |
| Quantity | 5 g |
| Synonym | Thiocyanic Acid Cobalt(2+) Salt, Cobalt Thiocyanate, Cobalt Dithiocyanate, Cobalt Isothiocyanate, Cobalt Rhodanide, Cobalt(2+) Thiocyanate, Cobalt(II) Thiocyanate, Cobaltous Thiocyanate, Isothiocyanic Acid Cobalt(2+) Salt |
| InChI Formula | InChI=1S/2CHNS.Co/c2*2-1-3;/h2*3H;/q;;+2/p-2 |
| SMILES | [Co+2].[S-]C#N.[S-]C#N |
| IUPAC Name | cobalt(2+);dithiocyanate |
| Molecular Weight (g/mol) | 175.1 |
| Formula Weight | 174.8835 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction