missing translation for 'onlineSavingsMsg'
Learn More

Cobalt(II) Thiocyanate, TRC

Product Code. 30349779
Click to view available options
Quantity:
50 g
Unit Size:
50g
This item is not returnable. View return policy

Product Code. 30349779

Brand: Toronto Research Chemicals TRCC63312550G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cobalt(II) Thiocyanate
CAS 3017-60-5
Molecular Formula Co (S C N)2
Quantity 50 g
Synonym Thiocyanic Acid Cobalt(2+) Salt, Cobalt Thiocyanate, Cobalt Dithiocyanate, Cobalt Isothiocyanate, Cobalt Rhodanide, Cobalt(2+) Thiocyanate, Cobalt(II) Thiocyanate, Cobaltous Thiocyanate, Isothiocyanic Acid Cobalt(2+) Salt
InChI Formula InChI=1S/2CHNS.Co/c2*2-1-3;/h2*3H;/q;;+2/p-2
SMILES [Co+2].[S-]C#N.[S-]C#N
IUPAC Name cobalt(2+);dithiocyanate
Molecular Weight (g/mol) 175.1
Formula Weight 174.8835
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.