missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Nitrate, 6-Hydrate, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™

Product Code. 15567914 Shop All J.T.Baker Products
Change view
Click to view available options
Quantity:
500 g
Packaging:
Poly Bottle
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15567914 500 g Poly Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15567914 Supplier Avantor J.T.Baker Supplier No. 168001

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High quality chemicals for laboratory and specialized industrial use

Specifications

CAS 10026-22-9
Quantity 500 g
Molecular Formula CoH12N2O12
MDL Number MFCD00149647
Synonym cobalt nitrate hexahydrate, cobaltous nitrate hexahydrate, cobalt dinitrate hexahydrate, cobalt 2+ nitrate hexahydrate, cobaltous nitrate, hexahydrate, kobalt ii-nitrat-hexahydrat, nitric acid, cobalt 2+ salt, hexahydrate, cobalt ii nitrate hexahydrate
InChI Key QGUAJWGNOXCYJF-UHFFFAOYSA-N
SMILES [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.[Co+2]
IUPAC Name cobalt(2+);dinitrate;hexahydrate
Molecular Weight (g/mol) 291.03
PubChem CID 24821
ChEBI CHEBI:86214
Formula Weight 291.03
Grade BAKER ANALYZED™ ACS Reagent
Packaging Poly Bottle
Specific Gravity 1L = 1.87kg
Chemical Name or Material Cobalt Nitrate, 6-Hydrate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.