missing translation for 'onlineSavingsMsg'
Learn More

Cobalt Nitrate Hexahydrate, TRC

Product Code. 30136412
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
1g
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136412 2.5 g 2.5g
30010394 500 mg 500mg
30008975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30136412 Supplier Toronto Research Chemicals Supplier No. TRCC1002672.5G
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10026-22-9
Molecular Formula Co(NO3)2 . 6H2O
Molecular Weight (g/mol) 291.04
Synonym Cobalt(2+) Dinitrate Hexahydrate, Cobalt(2+) Nitrate Hexahydrate, Cobalt(II) Nitrate Hexahydrate, Cobaltous Dinitrate Hexahydrate, Cobaltous Nitrate Hexahydrate;
IUPAC Name cobalt(2+);dinitrate;hexahydrate
SMILES O.O.O.O.O.O.[Co+2].[O-][N+](=O)[O-].[O-][N+](=O)[O-]

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/Co.2NO3.6H2O/c;2*2-1(3)4;;;;;;/h;;;6*1H2/q+2;2*-1;;;;;;
Formula Weight 290.9722
Chemical Name or Material Cobalt Nitrate Hexahydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.