missing translation for 'onlineSavingsMsg'
Learn More

Cobaltocene, 98%

Product Code. 10025490 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10025490 1 g Glass bottle 1g
10350184 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10025490 Supplier Thermo Scientific Acros Supplier No. 405320010
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1277-43-6 | C10H10Co | 189.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1277-43-6
Molecular Formula C10H10Co
Molecular Weight (g/mol) 189.12
MDL Number MFCD00013749
InChI Key SNBRJOIYNQIWSZ-UHFFFAOYSA-N
Synonym Bis(cyclopentadienyl)cobalt
PubChem CID 24942376
IUPAC Name cobalt(2+);cyclopenta-1,3-diene
SMILES C1C=CC=[C-]1.C1C=CC=[C-]1.[Co+2]

Specifications

Melting Point 173°C
Color Black
Elemental Analysis 97.5% min.
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 1 g
Solubility Information Solubility in water: insoluble.,soluble in hydrocarbons
Formula Weight 189.12
Percent Purity 98%
Physical Form Crystals
Chemical Name or Material Cobaltocene
Show More Show Less
compliance-icons
Product Identifier
  • Cobaltocene
Signal Word
  • Warning
Hazard Category
  • Flammable solid Category 2
Hazard Statement
  • H228-Flammable solid.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.